Learn More
Evocalcet, MedChemExpress
MedChemExpress Evocalcet has an activating effect on calcium sensing receptor (CaSR) extracted from patent WO 2017061621 A1, compound A.
£100.43 - £566.28
Specifications
Chemical Name or Material | Evocalcet |
---|---|
CAS | 870964-67-3 |
Color | Off-White |
Purity Grade Notes | Research |
Molecular Formula | C24H26N2O2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16487418
|
Medchem Express
HY-17613 |
10 mg |
£151.25
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16479897
|
Medchem Express
HY-17613 |
5 mg |
£100.43
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16453447
|
Medchem Express
HY-17613 |
50 mg |
£566.28
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Evocalcet has an activating effect on calcium sensing receptor (CaSR) extracted from patent WO 2017061621 A1, compound A.Specifications
Evocalcet | |
Off-White | |
C24H26N2O2 | |
DMSO : 30 mg/mL (80.11 mM; Need ultrasonic and warming) | |
374.48 | |
99.05% | |
Solid |
870964-67-3 | |
Research | |
KHK7580 | |
O=C(O)CC1=CC=C(N2C[C@@H](N[C@@H](C3=C4C=CC=CC4=CC=C3)C)CC2)C=C1 | |
374.48 | |
Research | |
Metabolism-protein/nucleotide metabolism |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.