missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Evolution™ Pro UV-Vis Spectrophotometers

Thermo Scientific™ Evolution™ UV-Vis Spectrophotometers combine reliable and versatile hardware with easy-to-learn, easy-to-use Thermo Scientific™ Insight™ Pro Software. Together they will meet the diverse needs your organization requires from an analytical tool.

POA - £14990.00

Specifications

Product Line Evolution Pro
Spectral Bandwidth 0.5, 1.0, 1.5, 2.0, 4.0 nm Selectable
View More Specs

Products 6
Product Code Brand Description Spectral Bandwidth Price Quantity & Availability  
Product Code Brand Description Spectral Bandwidth Price Quantity & Availability  
17319263
View Documents Promotion Details
Thermo Scientific™
840-340200
Evolution Pro UV-Vis Spectrophotometer 0.5, 1.0, 1.5, 2.0, 4.0 nm Selectable
£13955.00
Each
Please to purchase this item. Need a web account? Register with us today!
17309263
View Documents
Thermo Scientific™
840-340100
Evolution Pro UV-Vis Spectrophotometer with Mercury lamp 0.5, 1.0, 1.5, 2.0, 4.0 nm Selectable
£14990.00
Each
Please to purchase this item. Need a web account? Register with us today!
17405613
View Documents Promotion Details
Thermo Scientific™
912A1112
Evolution Pro Pharma Value Bundle include:
- Evolution Pro UV-Vis Spectrometer
- INSIGHT Pro with Security Software
- Validator for Insight Pro A4 Paper Version
- Qualification Filter Kit for UV-Vis
0.5, 1.0, 1.5, 2.0, 4.0 nm Selectable
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

17485603
View Documents
Thermo Scientific™
840-372800
Evolution Pro Pharma Value Bundle include:
Evolution Pro w/Mercury Lamp UV-Vis Spectrometer
INSIGHT Pro with Security Software
Validator for Insight Pro A4 Paper Version
Qualification Filter Kit for UV-Vis
0.5, 1.0, 1.5, 2.0, 4.0 nm Selectable
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

17415613
View Documents
Thermo Scientific™
912A1113
Evolution Pro USP/EP Standards Pharma Bundle include:
- Evolution Pro UV-Vis Spectrometer
- INSIGHT Pro with Security Software
- Validator for Insight Pro A4 Paper Version
- USP and EP UV Standards Set
- USP and EP Visible Standards Set
0.5, 1.0, 1.5, 2.0, 4.0 nm Selectable
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

17495603
View Documents
Thermo Scientific™
840-372900
Evolution Pro USP/EP Standards Pharma Bundle include:
Evolution Pro w/ Mercury Lamp UV-Vis Spectrometer
INSIGHT Pro with Security Software
Validator for Insight Pro A4 Paper Version
USP and EP UV Standards Set
USP and EP Visible Standards Set
0.5, 1.0, 1.5, 2.0, 4.0 nm Selectable
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

Description
Show More
Specifications

Specifications

Evolution Pro
0.5, 1.0, 1.5, 2.0, 4.0 nm Selectable
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Evolution™ Pro UV-Vis Spectrophotometers

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.