Learn More
Thermo Scientific™ Evolution™ UV-Visible Spectrophotometers Smart Thermostatted 7-Cell Rotary Changer
Efficiently control temperature with a circulating water bath using the Thermo Scientific™ Evolution™ UV-Visible Spectrophotometers Smart Thermostatted 7-Cell Rotary Changer.
£1525.00 - £2630.00
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
12306583
|
Thermo Scientific™
840-214900 |
Smart Thermostatted Rotary 7-Cell Changer |
£2630.00
Each |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12730928
|
Thermo Scientific™
10011401 |
Temperature probe accessory for Smart accessories |
£1525.00
Each |
Estimated Shipment: 09-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Efficiently control temperature with a circulating water bath using the Thermo Scientific™ Evolution™ UV-Visible Spectrophotometers Smart Thermostatted 7-Cell Rotary Changer. Combined with a liquid recirculator, this cell changer provides control of sample temperature for up to 7 samples or standards.
- Easy-to-use sample positioning accessory for use with Thermo Scientific™ Evolution™ One/One Plus and Evolution™ Pro spectrophotometers.
- Accessory control fully integrated in Thermo Scientific™ INSIGHT™ Pro software.
- Automatic software recognition, smart alignment and serial number reporting
- Includes carousel and motor base.
- When placed in both sample and reference beams (Evolution 300 only), this cell changer allows comparison of up to 7 samples with 7 unique references.
- Monitor the temperature in a cell with Temperature probe accessory for Smart accessories, catalog number 0011401.
- When deploying this accessory in a regulated or QC environment, a full-system qualification package is available to assure compliance of instrument and accessory as a complete unit.