Learn More
EVOS™ EVOS™ 100X Oil Objective, achromat, coverslip-corrected
For general applications and image with different magnification
Brand: EVOS™ AMPFOP100
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Coverslip-corrected objective that is optimized for imaging through #1.5 coverslips
- Offers optical performance from visible light to near infrared light
- Meets needs across spectrum of magnifications and optical specifications
Specifications
Objective Lens | |
EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System | |
0.17 mm | |
100X | |
1 Ea. |
EVOS | |
Vessels, Slides, Cell Culture Dishes, Flasks, Microtiter Well Plates | |
Achromat | |
1.25 |
For Research Use Only. Not for use in diagnostic procedures.