missing translation for 'onlineSavingsMsg'
Learn More

EVOS™ 20X Objective, fluorite, coverslip-corrected

EVOS™ 20X Objective, fluorite, coverslip-corrected

Brand:  Invitrogen AMEP4698

Product Code. 11927315

  • £1934.00 / Each
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

This fluorite objective is ideal for fluorescence and demanding transmitted-light applications. This is a coverslip-corrected objective that is optimized for imaging through #1.5 coverslips. All EVOS™ objectives offer outstanding optical performance from visible light to near infrared light. The extensive choice of objectives satisfies needs across the spectrum of magnifications and optical specifications.

Additional characteristics of this EVOS™ objective:

  • Magnification: 20X
  • Numerical Aperture: 0.50
  • Working Distance: 2.5 mm

    Image Quality
    Microscope objectives may be the most important components of an optical microscope because they are responsible for primary image formation. Image quality is crucial to experimental success and a requirement for publication; EVOS™ objectives afford that quality across the visible spectrum to near infrared light. This performance results from years of lens manufacture perfection. EVOS™ objectives have the same or better numerical apertures as any other manufacturer's in the same class, and the broad selection means you have choices for your imaging requirements.

    Objective Classes

    Achromat objectives are perfect for general applications, with standard correction of color and focus.

    Fluorite objectives deliver excellent resolution and are made with higher numerical apertures than achromat objectives, resulting in brighter fluorescence signal and higher contrast imaging. The higher optical quality greatly reduces optical aberrations, and corrections for color and focus are at higher levels than achromat objectives. Fluorite objectives are ideally suited for fluorescence and demanding transmitted light applications, where the higher contrast make them ideally suited for color imaging.

    Apochromat objectives are manufactured to the highest levels of resolution, fluorescence brightness, and contrast; chromatic aberrations are almost eliminated. They are recommended for the most demanding applications, particularly at magnifications of 60x and above. Apochromatic objectives are the best choice for the capture of color images in white light.

    Brightfield Contrast versus Phase Contrast Objectives
    Brightfield is the most basic form of light microscopy and is accomplished by sample absorption of light. A higher density area in a sample will absorb more light, thus increasing contrast in those areas.

    Phase contrast objectives are most useful for hard to see, translucent specimens. This method of contrast is accomplished by converting phase shifts, caused by light passing through a translucent specimen, into brightness changes (i.e., contrast).

    Long Working Distance versus Coverslip-Corrected Objectives
    Long working distance (LWD) objectives are optimized for use through vessels with a nominal wall thickness of 0.9-1.5 mm. This includes vessels commonly used in cell culture and cell-based assays, such as slides, cell culture dishes and flasks, microtiter well plates, etc. Coverslip-corrected objectives are optimized for use through #1.5 coverslips (thickness approximately 0.17 mm). These objectives have a higher magnification/NA ratio and provide higher resolution compared to LWD objectives.

    For additional choices, visit the EVOS™ Objectives selection guide
    Explore the entire EVOS™ line of imaging systems and accessories
    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    Objective Lens
    Objective Lens, 20X, Plan Fluorite, Coverslip Corrected, 0.5 Numerical Aperture, 2.5mm Working Distance, This Fluorite Objective is Ideal for Fluorescence and Demanding Transmitted Light Applications
    EVOS™ FL auto imaging system, EVOS™ FL color imaging system, EVOS™ FL imaging system, EVOS™ XL core imaging system, EVOS™ XL imaging system
    1 Each
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

    For Research Use Only. Not for use in diagnostic procedures.