Learn More
EVOS™ EVOS™ Vessel Holder for Automated Stage, two 25 x 75 mm slides
For use with automated stage of the EVOS™ FL Auto Imaging System
Brand:  EVOS™ AMEPVH021
Product Code. 15230211
  • £405.00 / Each
Estimated Shipment: 22-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Allows user to utilize different types of vessels for imaging
  • Fits microscope slide, cell culture flask or dish, or micro-well plate to stage of EVOS imaging systems for increased precision in sample alignment
  • Includes easy-to-change stage plates and vessel holders for this instrument platform
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Vessel Holder
EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
2 25 x 75 mm Microscope Slides
EVOS
2 Slides
1 each