Learn More
EVOS™ EVOS™ FL Auto Master Plate for Large Vessels
Designed for use with EVOS™ FL Auto Imaging System
Brand: EVOS™ AMEPVH027
Code : NEW
Additional Details : Weight : 2.00000kg
Description
- Allows user to utilize different types of vessels for imaging of cells grown in large cell culture flasks and dishes
- Suitable for T150 and larger tissue culture flasks
- Has easy-to-change plates and holders available for this instrument platform, for optimal precision in sample alignment
Specifications
Master Plate | |
EVOS | |
EVOS™ FL Auto Imaging System | |
Large Tissue Culture Flask, T150 Vessel |
For Research Use Only. Not for use in diagnostic procedures.