Learn More
EVOS™ EVOS™ FL Condenser Slider Block
Blocks fluorescent light from being reflected by condenser
Brand: EVOS™ AMEP4688
This item is not returnable.
View return policy
Description
- Improves image quality
- Serves as alternative to light shield box for sample access during fluorescence imaging in dark environments
- Compatible with EVOS FL , FL Color, and FL Auto imaging systems
Specifications
Fl Condenser Slider Block | |
EVOS™ FL Color Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System | |
1 Ea. |
EVOS | |
Condensing Optics, Hard-Coated Filters, LED |
For Research Use Only. Not for use in diagnostic procedures.