missing translation for 'onlineSavingsMsg'
Learn More

EVOS™ EVOS™ 4X Objective, fluorite, LWD

Components manufactured to ensure publication-quality images

Specifications

For Use With (Equipment) EVOS™ digital microscope
View More Specs

Description

Description

RMS-threaded objectives with 45mm parfocal distance

Specifications

Specifications

EVOS™ digital microscope
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.