Learn More
EVOS™ Onstage Incubator Hose
EVOS™ Onstage Incubator Hose
Brand: Invitrogen AMEP4728
Code : NEW
Additional Details : Weight : 2.00000kg
Description
The EVOS™ Onstage Incubator Hose connects the Control Unit and the Stagetop Environmental Chamber units of the EVOS™ Onstage Incubator and helps precisely control the temperature. Even small temperature shifts can have profound effects on dynamic cellular events. When performing live cell imaging studies, it is essential that the physiological and biological processes under investigation are not altered in any way. The ability to maintain temperature control with a precision of 0.1°C helps maintain cell viability and function and helps reduce the risk of biological temperature-related artifacts.Explore the entire EVOS™ line of imaging systems and accessories.
Specifications
Onstage Accessory | |
EVOS™ Onstage Incubator Hose, Onstage Accessory, Connects the Control Unit and the Stagetop Environmental Chamber Unit, Helps Precisely Control the CO2 and O2 Levels, Use With EVOS™ FL Auto Imaging System and Cellular Imaging | |
EVOS™ FL Auto Imaging System | |
1 Each |
For Research Use Only. Not for use in diagnostic procedures.