Learn More
Invitrogen™ EVOS™ Onstage Incubator (OSI-2) Oxygen Sensor Replacement Kit
The EVOS Onstage Incubator (OSI-2) Oxygen Sensor Replacement Kit contains everything needed for the self-installation of a new OSI-2 O2 sensor.
Brand: Invitrogen™ AMEP4977
Additional Details : Weight : 0.90700kg
Description
The EVOS Onstage Incubator (OSI-2) Oxygen Sensor Replacement Kit contains everything needed for the self-installation of a new OSI-2 O2 sensor. It is recommended that the OSI-2 O2 sensor be replaced every four years, particularly if the OSI-2 is regularly used for hypoxic experiments. The EVOS M5000 and EVOS M7000 user interface is designed to alert users when a replacement is recommended based upon the amount of time that has passed since the last O2 sensor replacement.
Specifications
EVOS | |
EVOS™ OSI-2 Oxygen Sensor Replacement Kit | |
EVOS™ Onstage Incubator (OSI-2) | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.