Learn More
Thermo Scientific™ EVOS™ Power Cord, Type C (Europe)
This power cord is for use with EVOS XL, FL , FL Color, and FL Auto systems in countries with Type C power plugs.
Brand: Thermo Scientific™ AMEP4646
Code : NEW
Additional Details : Weight : 1.00000kg
Description
This power cord is for use with EVOS XL, FL , FL Color, and FL Auto systems in countries with Type C power plugs. The Type C electrical plug (Europlug) is used throughout continental Europe, parts of the Middle East, and much of Africa, South America, Russia, and the former Soviet republics.
Specifications
Power Cord | |
Type C (Electrical Plug), Type C (Power Plug) | |
1 Ea. |
EVOS | |
EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System |