Learn More
Thermo Scientific™ EVOS™ Power Cord, Type G (United Kingdom)
This power cord is for use with EVOS XL, FL , FL Color, and FL Auto systems in countries or regions with Type G power plugs.
Brand: Thermo Scientific™ AMEP4645
Code : NEW
Additional Details : Weight : 0.10000kg
Description
This power cord is for use with EVOS XL, FL , FL Color, and FL Auto systems in countries or regions with Type G power plugs. The Type G electrical plug is used in the UK, Ireland, Hong Kong, Singapore, and other parts of Africa and Asia.
Specifications
Power Cord | |
Type G (Electrical Plug), Type G (Power Plug) | |
1 Ea. |
EVOS | |
EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System |