Learn More
EVOS™ EVOS™ Vessel Holder, ibidi 50 mm Petri dishes
For flasks, trays and petri dishes
Brand:  EVOS™ AMEPVH014
Code : NEW
Additional Details : Weight : 0.01000kg
Product Code. 11997325
  • £126.00 / Each
Estimated Shipment: 12-08-2024
to see stock.
Add to basket
Description
Description
Broad range of holders helps maximize the flexibility of the EVOS and Micromaster™ inverted microscopes.
Specifications
Specifications
Vessel Holder
EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
2 50 mm ibidi Petri Dishes
EVOS
Ibidi 50 mm Petri dishes
1 Holder