Learn More
EVOS™ EVOS™ Vessel Holders Pack
Provides snug and secure fit of microscope slide, cell culture flask or dish, or micro-well plate to the stage of the EVOS imaging systems
Brand:  EVOS™ AMEP4619
Code : NEW
Additional Details : Weight : 0.01000kg
Product Code. 11530217
  • £852.00 / Each
Estimated Shipment: 14-08-2024
to see stock.
Description
Description
  • Provides increased precision in sample alignment
  • Easily switchable vessel holders for convenient and easy imaging of a range of samples
  • Includes 10 most popular EVOS vessel holders
  • For use with: EVOS XL Core Imaging System, EVOS FL Auto Imaging System, EVOS XL Imaging System, EVOS FL Imaging System, EVOS FL Color Imaging System
Specifications
Specifications
Vessel Holder
EVOS
EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
T-25 flask