missing translation for 'onlineSavingsMsg'
Learn More

CleanSpace™ EX Half Masks for PAPR System

EX Half Masks are specifically designed to clip onto and complete the CleanSpace™ EX PAPR system.

Brand:  CleanSpace™ PAF-0064

 View more versions of this product

Product Code. 15920072

  • £137.00 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Used with the revolutionary CleanSpace™ PAPR: lightweight, no hoses or belts
  • Half masks available in 3 sizes to facilitate a secure seal and a comfortable fit
  • Materials: Soft silicone cushion, nylon clips and polycarbonate (exhalation valve)
  • Mask Fit Test adaptor available for TSI PortaCount fit testing
  • Easy removal for cleaning
  • Transparent and facilitates communication
  • Half masks clip quickly onto the power units and are easily unclipped for cleaning
  • Soft silicone that readily moulds to the face and does not cause allergic dermatitis
  • Treated with an electrically conductive coating and have been approved for the use in Intrinsically safe environments
Specifications

Specifications

AS/NZS1716: 2012
Clear
Small
PAF-1060
PAF-0064
170 x 130 x 70 mm
EN 12942
PAPR Half Mask
Mining, welding, manufacturing, smelting, construction, recycling plants, emergency services, agriculture, processing plants, grinding
IP66/67
Store at -10°C to 55°C
125 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers