missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva ExcelGel 12.5%, Tris-Acetate, Protein Gel, 1-well
Separation Range: 10 to 120 kd; Separation Type: Denaturing; For Use With: Multiphor II Electrophoresis Unit, ExcelGel SDS BufferStrips
Brand: Cytiva 17-6002-21
This item is not returnable.
View return policy
Description
Order Info
Order IPG strips separately.
Specifications
Polyester | |
12.5% | |
Tris-Acetate | |
Protein | |
Denaturing | |
SDS-PAGE | |
6/Pk. | |
2° to 8°C | |
4.3 in. | |
11 cm | |
9.8 in. | |
25 cm |
Multiphor II Unit, ExcelGel SDS BufferStrips | |
0.5 mm | |
Homogeneous Page | |
10 to 120 kDa | |
1-well | |
Polyacrylamide | |
77mm, 12.5/2% T/% C | |
4.3 in. | |
250 mm | |
9.8 in. | |
110 mm |