Learn More
Cytiva ExcelGel 15%, Tris-Acetate, plus Ethylene Glycol, 0.5 mm, Protein Gel, 25-well
Separation Range: 3 to 8 kd; Separation Type: Denaturing; For Use With: Immobiline DryStrip Gels
Brand: Cytiva 80-1262-01
Code : GE
Additional Details : Weight : 0.42200kg
Description
Three homogeneous and one gradient polyacrylamide gel for horizontal electrophoresis of SDS denatured proteins; also for separation of nucleic acids in the base pair range 50-1500
- Homogeneous 7.5, 12.5, and 15% gels have 25 preformed sample wells for sample volumes from up to 17μL for quick, easy sample application
- Gradient gel uses sample application strips (26 samples up to 40μL, 52 samples up to 20μL), paper pieces, or sample droplets; recommended for use with Immobiline DryStrip gels
- ExcelGel SDS Homogeneous 15% is also designed for separation of peptides
Specifications
Polyester | |
15% | |
Tris-Acetate, plus Ethylene Glycol | |
Protein | |
Denaturing | |
Analytical Chemistry and Research, Research Horizontal Electrophoresis of SDS Denatured Proteins | |
71.08 | |
7.5% T/3% C | |
0.019 in. | |
4.3 in. | |
11 cm | |
9.8 in. | |
25 cm |
Immobiline DryStrip Gels | |
0.5 mm | |
Homogeneous Page | |
3 to 8 kDa | |
25-well | |
Polyester Film | |
6/Pk. | |
2° to 8°C | |
4.3 in. | |
110 mm | |
9.8 in. | |
25 cm |