Learn More
Cytiva ExcelGel DNA Analysis Kit
Provide fast, easy, high resolution and reproducible manual analysis of DNA fragments in polyacrylamide gels
Brand: Cytiva 17-1198-07
Code : GH
Additional Details : Weight : 0.54400kg
Description
Kits are intended for use with Multiphor™ II Flatbed Electrophoresis System. Provide more reliable results than those obtained with agarose electrophoresis: optimal separation range is 50-500 base pairs (bp), with resolution of 6-8 bp in the 100–200 bp range.
- Achieve high throughput with precast gels with 48 preformed 5-7μL sample wells
- DNA fragments can be visualized in about 120 minutes
- Silver staining following separations yields high senstivity and permanent documentation of results
- Gel dimensions: 250 x 110 x 0.5 mm
- Gels offers long shelf life--up to 24 months
ExcelGel™ (12.5% gel) Kit
- Offers the convenience of a ready-to-use gel; no need to handle buffer and rehydration solutions
- Used in a variety of DNA techniques: SSCP, RAPD, and VNTR
- Includes five ExcelGel 48S gels and 12 buffer strips, and instructions
Specifications
Multiwell | |
100 to 1500 bp | |
Multiphor™ II Flatbed Electrophoresis System | |
ExcelGel 48S |
Precast Gel | |
48-well | |
Five ExcelGel 48S gels with 48 5 to 7μL wells, six anode Buffer strips and six cathode Buffer strips and manual | |
1 Each |
Safety and Handling
- Danger
Hazard Category
- Germ cell mutagenicity Category 1B
- Carcinogenicity Category 1B
Hazard Statement
- H340-May cause genetic defects.
- H350-May cause cancer.
- EUH208-Contains Acrylamide. May produce an allergic reaction.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P405-Store locked up.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P501-Dispose of contents/container to in accordance withall local, regional,national and international regulations.
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance withall local, regional,national and international regulations.
Supplemental information
- MIXTURE LIST-Contains : Acrylamide