Learn More
Cytiva ExcelGel SDS Buffer Strips
Semi dry precast buffer strips for horizontal SDS electrophoresis.  Cytiva Lifescience™ ExcelGel SDS Buffer Strips are designed for use with ExcelGel Precast Gels.
Brand:  Cytiva 17-1342-01
Code : GE
Additional Details : Weight : 0.29700kg
Product Code. 11515015
  • £120.65 / Pack of 6
Estimated Shipment: 18-07-2024
to see stock.
Description
Description
  • Designed for use with ExcelGel Precast Gels for SDS-PAGE
  • Include strips for both anode and cathode
Specifications
Specifications
Multiphor II Flatbed Electrophoresis System
Acrylamide 0.55%
6 Pack
ExcelGel SDS Buffer strips
SDS-PAGE
Cool, dry place
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Danger
Hazard Category
  • Germ cell mutagenicity Category 1B
  • Carcinogenicity Category 1B
Hazard Statement
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • EUH208-Contains Acrylamide. May produce an allergic reaction.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Acrylamide
  • UNKNOWN ACUTE-75 per cent of the mixture consists of component(s) of unknown acute toxicity.