missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Excelsior™ AS Tissue Processor Accessories
Maintain the Epredia™ Excelsior™ AS Tissue Processor with Epredia™ Excelsior AS Tissue Processor Accessories.
Brand: Epredia™ A78410026
113.90 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
For use with Shandon Excelsior ES Tissue ProcessorSpecifications
Storage Bottle | |
Excelsior AS Tissue Processor | |
5 L, 6/Pk. |