Learn More
Epredia™ Excelsior™ AS Tissue Processor Accessories
Maintain the Epredia™ Excelsior™ AS Tissue Processor with Epredia™ Excelsior AS Tissue Processor Accessories.
Brand: Epredia™ A82310038
598.08 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
Product Features:- Microwaveable design allowing pre-processing of tissue
- Organized capacity of 300–cassettes in six basket-halves
Ordering Alerts:
Basket halves only match with each other but can be mixed with 222 cassette baskets or SecureSette Excelsior ES baskets
Includes:
Six 50-cassette basket halves and user guide and loading instructions
Compatible with:
Excelsior ES Tissue Processor
Specifications
Organized Basket, 6 Sections | |
Excelsior AS Tissue Processor | |
300 Cassettes Kit | |
Ambient |