missing translation for 'onlineSavingsMsg'
Learn More

SI Analytics™ Exchange Unit

SI Analytics™ Exchange Unit is a unit that includes an integrated chip for reagent dat, tubes, a titration tip, and more.

Brand:  SI Analytics™ 285220300

 View more versions of this product

Product Code. 13476979

  • £715.00 / Each
Estimated Shipment: 14-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Includes integrated chip for reagent data
  • Comes with a brown glass bottle for titrant
  • GL 45
  • S 40 bottle adapter
  • Includes tubes, drip tubes, and titration tip
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Brown glass bottle for titrant
Exchange Unit
5 mL
GL 45
Integrated chip for reagent data
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
SI Analytics™ Exchange Unit > Capacity: 5mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.