Learn More
Eppendorf™ Exchangeable Thermoblock
Product Code. 10439472
Click to view available options
For Use With (Equipment):
24 Reaction Vessels 0.5mL with Work Rack and Cool Pack 0°C
24 Reaction Vessels 1.5mL with Work Rack and Cool Pack 0°C
Accessories for ThermoStat plus/thermoMixer plus
  • Compact, bench top thermostat with thermoblocks for 0.2mL, 0.5mL, 1.0mL, 2.0mL tubes and microtitre plates designed to provide a high degree of flexibility for a multitude of applications
  • Programmable temperature and run time for special applications
  • GLP compliant
  • RS232 interface
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 24 Reaction Vessels 1.5mL with Work Rack and Cool Pack 0°C
Holds 1.5mL tubes
No. of Places 24
Type Block