Learn More
PSF Progressive Safety™ Executive Black Shoe
Water resistant black leather shoe
Brand:  PSF Progressive Safety™ 806SM-6
Code : 91
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12827583
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Four eyelet fasten
  • Antistatic
  • Steel toecap and midsole
  • Dual density rubber sole
  • SRC slip resistant
Specifications
Specifications
6