missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ ExoSAP-IT Express PCR Product Cleanup with Tracking Dye
Our fastest PCR clean-up method - just 5 minutes to superior sequencing results. Now available with optional tracking dye. It is necessary to remove excess primers and unincorporated nucleotides from the PCR reaction prior to sequencing.
£622.00 - £2410.00
Specifications
Content And Storage | Shipped on dry ice. Store at -20°C. |
---|---|
Product Line | ExoSAP-IT™ |
Protein Form | Recombinant |
Research Category | Genetic Diseases |
Shipping Condition | Dry Ice |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
15889916
|
Applied Biosystems™
A35004 |
500 Reactions |
£622.00
500 reactions |
Estimated Shipment: 24-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15899916
|
Applied Biosystems™
A35005 |
1920 reactions |
£2410.00
Each |
Estimated Shipment: 25-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- 5 minute protocol: Fastest turnaround time to results
- One-tube, one-step PCR clean-up: Add ExoSAP-IT Express directly to PCR product
- Novel enzyme technology: Irreversibly inactivated in just one minute at 80°C
- Conserve PCR samples: 100% recovery of PCR products, regardless of amplicon length
- Eliminate spin columns or beads: Significantly decreases time and errors while increasing yield
- Compatible: No interference in downstream applications
- Scalable: Treat a wide range of volumes
- Multiple formats: Single tube, 8 tube strip, and 96-well plate formats offer flexibility from manual pipetting to automated workflows
- Go green: Generate less waste with a single-tube solution to PCR clean-up
- Optional tracking dye: Never lose track of reagent addition during pipetting
Specifications
Shipped on dry ice. Store at -20°C. | |
Recombinant | |
Dry Ice |
ExoSAP-IT™ | |
Genetic Diseases |
For Research Use Only. Not for use in diagnostic procedures.