missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ ExoSAP-IT™ PCR Product Cleanup Reagent
ExoSAP-IT™ PCR Product Clean-up Reagent is used for enzymatic clean-up of amplified PCR product. It hydrolyses excess primers and nucleotides in a single step.
£117.00 - £2665.00
Specifications
Content And Storage | Shipped on dry ice. Store at -20°C. |
---|---|
Sample Type | PCR Products |
For Use With (Application) | PCR Purification |
Product Line | ExoSAP-IT™ |
Protein Form | Recombinant |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
15839906
|
Applied Biosystems™
78205.10.ML |
5000 Reactions | N/A | ||||||
15829906
|
Applied Biosystems™
78202.4X.1.ML |
2000 Reactions | N/A | ||||||
14880652
|
Applied Biosystems™
78201.1.ML |
500 Reactions |
£450.00
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12664537
|
Applied Biosystems™
78200.200.UL |
100 Reactions |
£117.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Remove excess primers and unincorporated nucleotides enzymatically
- Prepare DNA sequencing samples with one pipetting step
- Conserve precious PCR products with up to 100% recovery
- Minimize column clean-up and sample loss
Specifications
Shipped on dry ice. Store at -20°C. | |
PCR Purification | |
Recombinant | |
Dry Ice |
PCR Products | |
ExoSAP-IT™ | |
Genetic Diseases |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Applied Biosystems™ ExoSAP-IT™ PCR Product Cleanup Reagent