missing translation for 'onlineSavingsMsg'
Learn More

Exosome Component 9 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications

Brand:  Novus Biologicals NBP1-71702

 View more versions of this product

Product Code. 18701354

  • £395.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Exosome Component 9 Polyclonal specifically detects Exosome Component 9 in Human, Mouse, Chicken samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Exosome Component 9
Polyclonal
Unconjugated
PBS and 30% Glycerol with 0.05% Sodium Azide
Autoantigen PM/Scl 1, EC 3.1.13, exosome component 9Polymyositis/scleroderma autoantigen 75 kDa, p6, PM/Scl-75P75 polymyositis-scleroderma overlap syndrome-associated autoantigen, PMSCL175kD, Polymyositis/scleroderma autoantigen 1, polymyositis/scleroderma autoantigen 1 (75kD), polymyositis/scleroderma autoantigen 1, 75kDa
Rabbit
Affinity Purified
RUO
5393
Human, Mouse, Chicken
IgG
Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
0.6 mg/mL
Western Blot 1:5000, Simple Western 1:1000, Chromatin Immunoprecipitation reported in scientific literature (PMID 27543448), Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation reported by customer review, Immunohistochemistry-Paraffin 1:50-1:100, Chromatin Immunoprecipitation (ChIP)
Q06265, Q9JHI7
EXOSC9
A genomic peptide made to an internal region of human Exosome Component 9 (within residues 250-439). [Swiss-Prot Q06265]
0.1 mL
Primary
Human, Mouse and Chicken. Immunogen has 83% identity to rat and 90% identity to bovine.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only