missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Exosome Standards (LnCAP cell line)
Shop All Bio Techne Products
Click to view available options
Quantity:
2 x 100 μg Vials
2 x 30 μg Vials
6 x 100 μg Vials
6 x 30 μg Vials
Unit Size:
Pack of 2
Pack of 6
Specifications
Specifications
| Content And Storage | Store at 4°C. After reconstitution store at -70°C. |
| Concentration | LYOPH |
| Quantity | 2 x 100 μg Vials |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction