missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Exosome Standards (U87 MG)

Highly pure, lyophilized exosome standards with superior stability, optimal for multiple applications

£322.00 - £1040.00

Specifications

Content And Storage Store at 4°C. After reconstitution store at -70°C.
Concentration LYOPH
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18653820
View Documents
Novus Biologicals™
NBP2-49844-60ug
2 x 30 μg Vials
£322.00
Pack of 2
Please to purchase this item. Need a web account? Register with us today!
18616461
View Documents
Novus Biologicals™
NBP2-49844-200ug
2 x 100 μg Vials
£411.00
Pack of 2
Please to purchase this item. Need a web account? Register with us today!
18674850
View Documents
Novus Biologicals™
NBP2-49844-180ug
6 x 30 μg Vials
£835.00
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
18673320
View Documents
Novus Biologicals™
NBP2-49844-600ug
6 x 100 μg Vials
£1040.00
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Store at 4°C. After reconstitution store at -70°C.
LYOPH
Videos
SDS
Documents

Documents

Product Certifications