Learn More
Gibco™ Expi293™ Expression Medium
Description
Expi293 Expression Medium is a chemically defined, serum-free, protein-free medium for growth and transfection of suspension-adapted human embryonic kidney (HEK) 293 cells. It is specifically designed as a core component to support high density culture of Expi293F cells in the Expi293 Expression System for scalable transient protein expression.
- Supports growth of suspension 293 cultures to densities of over 1.5 x 10^7 cells/mL
- Transfection compatible serum-free suspension culture medium enables transfection efficiencies of greater than 70% using ExpiFectamine transfection reagent
- Enables sustained, high level expression of high density transiently transfected cultures, achieving yields of up to 1 gram per liter of recombinant protein
- Supports growth and transfection of 293 cells in culture formats of less than 1 mL in multiwell plates to greater than 10 L in disposable bioreactors
- Does not contain phenol red
Expi293 Expression Medium is formulated with GlutaMAX-I reagent. It is ready to use, with no supplementation required. The chemically defined formulation results in high reproducibility and lot-to-lot reliability. Expi293 Expression Medium contains no human or animal-origin components. The medium is not recommended for adherent cell cultures.
Specifications
Specifications
| Cell Line | Expi293F™ |
| Classification | Animal Origin-free, Chemically-defined, Protein-free, Serum-free |
| Form | Liquid |
| Product Type | Expression Medium |
| With Additives | GlutaMAX |
| Without Additives | No Phenol Red |
| Product Line | Expi293 |
| Quantity | 20 L |
| Content And Storage | One 20L Universal Bag Store in the dark at 2-8°C. |
For Research use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.