missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Expi293™ Expression Medium

Expi293™ Expression Medium

Brand:  Gibco™ A1435103

2345.25 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.



Code : NEW

Additional Details : Weight : 10.00000kg

 View more versions of this product

Product Code. 15490694

  • £2650.00 / 10 L
Estimated Shipment: 17-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Expi293™ Expression Medium is a chemically defined, serum-free, protein-free medium for growth and transfection of suspension-adapted human embryonic kidney (HEK) 293 cells. It is specifically designed as a core component to support high density culture of Expi293F™ cells in the Expi293™ Expression System for scalable transient protein expression.

  • Supports growth of suspension 293 cultures to densities of over 1.5 x 10^7 cells/mL
  • Transfection compatible serum-free suspension culture medium enables transfection efficiencies of greater than 70% using ExpiFectamine™ transfection reagent
  • Enables sustained, high level expression of high density transiently transfected cultures, achieving yields of up to 1 gram per liter of recombinant protein
  • Supports growth and transfection of 293 cells in culture formats of less than 1 mL in multiwell plates to greater than 10L in disposable bioreactors
  • Does not contain phenol red

    Expi293™ Expression Medium is formulated with GlutaMAX™-I reagent. It is ready to use, with no supplementation required. The chemically defined formulation results in high reproducibility and lot-to-lot reliability. Expi293™ Expression Medium contains no human or animal-origin components. The medium is not recommended for adherent cell cultures.
  • Specifications

    Specifications

    One 10L Universal Bag
    Store in the dark at 2-8°C.
    Animal Origin-free, Chemically-defined, Protein-free, Serum-free
    Expression Medium
    No Phenol Red
    10 L
    Expi293F™
    Liquid
    GlutaMAX
    Expi293™
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    For Research use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.