Learn More
Gibco™ Expi293™ Met (–) Expression Medium
Expi293 Met (–) Expression Medium is a cGMP-manufactured, chemically defined, serum-free, protein-free, animal origin-free, methionine-free medium for growth and transfection of Expi293F cells.
Brand: Gibco™ A4096701
390.22 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 1.13400kg
Description
- The medium is part of the Expi293 platform for rapid, high-yield transient protein expression from mammalian HEK293 cells
- Designed for use in combination with the ExpiFectamine 293 Met (–) Transfection Kit for applications where labelling of methionine residues in proteins is desired
- Note: the medium will not support cell growth or expression unless supplemented with methionine
Features:
- Supports growth of suspension 293 cultures to densities of over 1.5 x 107 cells/mL
- Equivalent performance to Expi293 Expression Medium
- Does not contain methionine
- Does not contain phenol red
- Formulated with GlutaMAX I Supplement
- Supplementation with methionine required
Specifications
293 Expression Medium | |
AOF, Chemically Defined, Methionine-Free, Protein-Free, Serum Free | |
Expi293™ |
1 L Expi293 Met (–) Expression Medium, store at 2–8°C (protect from light) |
|
Room Temperature | |
Liquid |
For Research Use Only. Not for use in diagnostic procedures.