Learn More
Gibco™ Expi293™ Met (–) Protein Labeling Kit
Expi293 Met (-) Protein Labeling Kit includes Expi293 Met (-) Expression Medium and the ExpiFectamine 293 Met (-) Transfection Kit for rapid, high-yield transient protein expression from Expi293F cells for applications where labeling of methionine residues in proteins is desired
Brand: Gibco™ A41249
Additional Details : Weight : 1.46400kg
Description
Kit components are specifically formulated without methionine, allowing users to add labeled methionine of their choice
Features:
- Methionine-free medium and transfection enhancer formulations
- 2 to 10-fold higher protein yields compared to other transfection reagents used for high-density 293 cell culture
- Same transient transfection protocol as ExpiFectamine 293 Transfection Kit
- Scalable transfection from small to large scale while maintaining equivalent volumetric protein yields
Specifically designed to easily integrate with other components of the Expi293 Expression System, including Expi293F cells and Expi293 Expression Medium
Components included:
- 1 L Expi293 Met (–) Expression Medium
- 1 ExpiFectamine 293 Met (–) Transfection Kit sufficient to transfect 1 L of culture
Specifications
Protein Labeling Kit | |
|
|
Expi293F™ |
Protein Production | |
Approved for shipment at Room Temperature or on Wet or Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.