missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Expi293F™ Cells

Product Code. 13305554
Click to view available options
Quantity:
1 mL
6 x 1 mL
Unit Size:
6mL
Each
This item is not returnable. View return policy

Product Code. 13305554

Brand: Gibco™ A14527

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Expi293F human cells are derived from the 293 cell line and are a core component of the Expi293 Expression System.

Expi293F human cells are derived from the 293 cell line and are a core component of the Expi293 Expression System. They are maintained in suspension culture and will grow to high density in Expi293 Expression Medium. Expi293F cells are highly transfectable and generate superior protein yields compared to standard 293 cell lines in transient protein expression.

Expi293F Cells attributes include:

  • Suspension culture cells pre-adapted to Expi293 Expression Medium
  • Demonstrated higher protein expression levels than other 293 cells adapted to Expi293 Expression Medium
  • Exhibit more rapid cell growth and higher culture viabilities than standard suspension-adapted 293 cells
  • Consistent transient expression performance of Expi293F cells for over 30 passages beyond thaw
  • Also available from a cGMP bank (Cat. No. 100044202)

Frozen cells are supplied in a vial containing 1 mL of cells at 1 x 107 viable cells/mL in Expi293 Expression Medium and 10% DMSO. The cells should be thawed directly into Expi293 Expression Medium. The six-vial multipack eliminates the need to expand and rebank the cells in the lab.

Cells are typically grown in flasks on a shaker platform in a humidified, CO2 cell culture incubator. However, cultures are scalable from volumes of <1 mL in multiwell plates to >10 L in bioreactors. Protein harvest can be performed at 2–7 days post-transfection, depending on the protein expressed. For expression of recombinant antibodies, harvest is typically at 5–7 days post-transfection. There is no need for culture medium supplementation or batch feeding during expression.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 x 107 cells/mL, store in liquid nitrogen
Cell Line Expi293F
Cell Type Kidney (Embryonic)
Culture Type Suspension Cell Culture
Form Cryopreserved
Species Human
Media Recommendation Expi293™ Expression Medium
No. of Cells 1 x 107
Product Line Expi293
Product Type Expi293F Cells
Quantity 1 mL
Shipping Condition Dry Ice
Source Human
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.