Learn More
Gibco™ Expi293F™ GnTI- Cells
Expi293F GnTI- cell line is derived from engineered Expi293F cells that do not have N-acetylglucosaminyltransferase I (GnTI) activity and therefore lack complex N-glycans
Brand:  Gibco™ A39240
Additional Details : Weight : 0.00300kg
Product Code. 16346552
  • £1982.00 / Each
In Stock
to see stock.
Description
Description
Powerful tool for the high-yield expression of homogeneously glycosylated recombinant proteins.
Features:
  • Suspension, high-density culture in Expi293 Expression Medium
  • Homogeneous N-glycosylation of expressed proteins
  • Equivalent protein yields to parental Expi293F cells—up to 1 g/L
  • Significantly higher protein yields than other HEK293 GnTI- cell line alternatives
Frozen cells are supplied in a vial containing 1 mL of cells at 1 x 107 viable cells/mL in Expi293 Expression Medium and 10% DMSO. The cells should be thawed directly into Expi293 Expression Medium
Specifications
Specifications
1 vial Expi293F GnTI- Cells (1x107 cells/vial), store in vapor-phase liquid nitrogen
Expi293™
Dry Ice