missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ ExpiCHO™ Stable Production Medium

Gibco ExpiCHO Stable Production Medium (SPM) is specifically designed to simplify your cell line development process into production with minimal effort and a high degree of confidence.

Brand:  Gibco™ A3711001

153.90 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.


Product Code. 16294262

  • £167.00 / 1 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This chemically defined, protein-free, animal origin component-free formulation is fully optimized for large-scale fed-batch culture with ExpiCHO-S cells. ExpiCHO SPM supports an easy transition from transient to stable production, provides high titers without the need for additional medium optimization (see figures below), and works well with existing EfficientFeed C+ supplement.

ExpiCHO SPM is part of the ExpiCHO family, an end-to-end solution that empowers you to carry out your entire workflow from transient to stable expression without having to screen or optimize media and feeds.

Hassle-free scale-up
Simplify your process as you transition into stable clones with ExpiCHO SPM—no cell adaptation, no media optimization, no changes in cell doubling time and peak density. ExpiCHO SPM eases the transition using ExpiCHO-S cells and provides optimal performance with less need for fine-tuning from development scientists to get you to the next step faster. This medium supports high titers under standard fed-batch process conditions in conjunction with EfficientFeed C+ supplement. It is available in liquid format (this page) or in our AGT dry format from 10 L through 450 L equivalents with larger sizes available upon request.

ONE Solution
The ExpiCHO family includes products that are engineered to work seamlessly with each other. ExpiCHO SPM is a member of this family and allows expanded support of bioproduction from transient to stable protein expression. Look for ExpiCHO-branded protocols and methods to get optimal performance in the development process.

Members of the ExpiCHO family include:
• ExpiCHO Stable Production Medium (this page)
• ExpiCHO Expression Medium
• ExpiCHO-S cells
• Expifectamine CHO Transfection Kit

The power to streamline transfer to manufacturing
ExpiCHO SPM liquid format and AGT dry format demonstrate comparable cell growth and titer performance (see figure below). Therefore, scale-up and manufacturing using AGT dry format is simplified by knowing that the reconstituted AGT dry format will behave similarly to the liquid format used prior to scale-up. The AGT dry format is easy to reconstitute; just add water and it will automatically adjust pH and osmolality. This can help provide efficiencies in supply chain complexity, reconstitution complexity, and, therefore, overall expense.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in refrigerator (2°C to 8°C).
Animal Origin-free, Chemically-defined, Protein-free
Stable Production Medium (SPM)
ExpiCHO™
Room Temperature
ExpiCHO-S
Liquid
Bottle
1000 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.