Learn More
Gibco™ ExpiFectamine™ 293 Transfection Kit
Designed for transient transfection of high density cultures of HEK (human embryonic kidney) 293 cells.
Brand: Gibco™ A14526
30960.50 GBP valid until 2024-12-31
Use promo code "21880" to get your promotional price.
Code : NEW
Additional Details : Weight : 3.75000kg
Description
- The ExpiFectamine™ 293 Transfection Kit is a core component of the Expi293™ Expression System
- It is designed for transient transfection of high density cultures of HEK (human embryonic kidney) 293 cells
- The high-efficiency, cationic, lipid-based transfection reagent and transfection enhancers are designed to power the highest possible level of protein expression from Expi293F™ cells cultured in Expi293™ Expression Medium
- Achieves protein yields 2- to 10-fold higher than other transfection reagents used on high density 293 cell cultures
- Employs the same transient expression protocols typically used in current low-density 293 suspension culture systems to easily switch from low density systems to the high yield, high density Expi293™ Expression System
- Provides robust and reproducible transfection results, giving users greater confidence in their results
- Scale transfections for culture volumes of less than 1 mL to greater than 10 liters, while maintaining equivalent volumetric protein yields
- Store at 2°C to 8°C
Specifications
Expi293F™ | |
Five 10-L kits; sufficient for transfection of 50 L of culture Store at +2°C to +8°C |
|
Plasmid DNA | |
Transfection | |
Transfection Kit | |
For 50 L of culture |
Animal Origin-Free, Chemically Defined, Protein-Free, Serum-Free | |
Established Cell Lines | |
Lipid-based Transfection | |
ExpiFectamine™ | |
Cell Culture Grade | |
No |
For Research Use Only. Not for use in diagnostic procedures.