Learn More
Gibco™ ExpiFectamine™ CHO Transfection Kit
Description
The ExpiFectamine CHO Transfection Kit is a core component of the ExpiCHO Expression System Kit (Cat. No. A29133) and enables transient transfection of high-density cultures of Chinese hamster ovary (CHO) cells. The high-efficiency, cationic transfection reagent, coupled with an optimized transfection enhancer and culture feed, will help power the highest possible level of protein expression from ExpiCHO-S Cells (Cat. No. A29127) cultured in ExpiCHO Expression Medium (Cat. No. A2910001).
All components work together synergistically with expression yields of 1–3 g per L of transfected culture demonstrated for some antibody and non-antibody proteins.
Features of the ExpiFectamine CHO Transfection Kit include:
- Enables robust and reproducible transfection results, giving you greater confidence in your results and ability to use CHO cells from research to discovery
- Achieve >90% transfection efficiency of high-density suspension CHO cell culture, with matching transfection enhancer and culture feed to boost transfection performance and protein expression
- Uses one-third to one-half less plasmid DNA than other transfection reagents
- Scale growth and transfection in culture formats from <1 mL in deep well plates up to 5 L when using Nalgene angled-bottom shake flasks
- Flexible, reliable culture and transfection protocols with multiple options that readily fit into current transient expression workflows
The ExpiFectamine CHO Transfection Kit is specifically designed to work with ExpiCHO-S Cells in the ExpiCHO Expression System Kit. The transfection kit supplies the cationic ExpiFectamine CHO Transfection Reagent, ExpiFectamine CHO Transfection Enhancer, and ExpiCHO Feed. ExpiCHO-S Cells and ExpiCHO Expression Medium are available separately.
Specifications
Specifications
| Cell Line | ExpiCHO-S™, CHO-S, SFM Adapted |
| Classification | Animal Origin-free, Chemically-defined, Protein-free, Serum-free |
| Content And Storage | Kit includes: • ExpiFectamine™ CHO Reagent, 3.2 mL • ExpiFectamine™ CHO Enhancer, 6 mL • ExpiCHO Feed, 330 mL Sufficient for transfection of 1 L of culture Store in the dark at 2°C to 8°C. |
| Cell Type | Ovarian Cells |
| Format | Kit |
| Promoter | pcDNA™ 3.4 |
| Transfection Technique | Lipid-based Transfection |
| Product Line | ExpiFectamine |
| Product Type | CHO Transfection Kit |
| Quantity | For 1 L of culture |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.