Learn More
Koettermann™ Exploris™ Balance Bench
Designed to attenuate external vibrations.
Brand:  Koettermann™ 509-00001
Product Code. 14855421
  • £2185.00 / Each
Explore more special offers
Description
Description
  • Stand-alone balance
  • Attenuate external vibrations
  • Large mass, oscillation-damped mounting
  • Low-vibration balance platform
  • Balance block mechanically separated from the outer chassis of balance bench
  • Vibrations caused by impact are not transferred directly to balance block
Specifications
Specifications
60 x 90 x 75 cm
Balance Bench
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Koettermann™ Exploris™ Balance Bench > Balance Bench