missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Explosive hazard barrier tape, 250m x 75mm
Brand: Manutan A353850
Code : Z3
Additional Details : Weight : 1.00000kg
Description
- Explosive hazard
- Dimensions (L x W) : 250m x 75mm