missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EXPRESS One-Step SYBR™ GreenER™ Kit, with premixed ROX

£1054.00 - £4410.00

Specifications

Content And Storage EXPRESS One-Step SYBR™ GreenER™ SuperMix Kits are composed of 2 modules: EXPRESS SYBR™ GreenER™ RT module (containing SuperScript™ III Reverse Transcriptase and RNAseOUT™) and EXPRESS SYBR™ GreenER™ qPCR SuperMix with ROX premixed.

Store at -20°C upon receipt. Once thawed, the EXPRESS SYBR™ GreenER™ qPCR SuperMix can be stored for up to 1 month at +4·C. Product is guaranteed to be stable for 6 months when properly stored.
Detection Method SYBR
Format Tube
PCR Method 1-step RT-qPCR
Polymerase Taq DNA Polymerase
View More Specs

Products 2
Product Code Brand No. of Reactions Price Quantity & Availability  
Product Code Brand No. of Reactions Price Quantity & Availability  
10226113
View Documents Promotion Details
Invitrogen™
11790200
500 Reactions
£1054.00
Each
Please to purchase this item. Need a web account? Register with us today!
10124403
View Documents Promotion Details
Invitrogen™
1179001K
2500 Reactions
£4410.00
2500 reactions
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This kit comes with premixed ROX reference dye. EXPRESS One-Step SYBR GreenER SuperMix kits with a separate tube of ROX reference dye are also available.

  • Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase
  • The One-Step SuperScript™ III Reverse Transcriptase for one-step qRT-PCR
  • SYBR GreenER dye
  • UDG carryover protection - uracil-DNA-glycosylase (UDG)/dUTP to reduce carryover contamination
Specifications

Specifications

EXPRESS One-Step SYBR™ GreenER™ SuperMix Kits are composed of 2 modules: EXPRESS SYBR™ GreenER™ RT module (containing SuperScript™ III Reverse Transcriptase and RNAseOUT™) and EXPRESS SYBR™ GreenER™ qPCR SuperMix with ROX premixed.

Store at -20°C upon receipt. Once thawed, the EXPRESS SYBR™ GreenER™ qPCR SuperMix can be stored for up to 1 month at +4·C. Product is guaranteed to be stable for 6 months when properly stored.
Tube
Taq DNA Polymerase
7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System, Eppendorf™ MasterCycler™ RealPlex, StepOne™, Fast Mode, StepOne™, Standard Mode
Gene Expression
SYBR
1-step RT-qPCR
Fast or Standard
SYBR™ GreenER™
RNA
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.