Learn More
SYBR™ GreenER™ EXPRESS One-Step SYBR™ GreenER™ Kit, universal
EXPRESS One-Step SYBR GreenER kits are compatible with fast-cycling instruments for your SYBR-based qPCR experiments.
Brand:  Invitrogen™ 11780200
Code : 50
Additional Details : Weight : 0.13000kg
 View more versions of this product
Product Code. 10563454
  • £1094.00 / Each
Estimated Shipment: 26-07-2024
to see stock.
For Research Use Only. All usage must comply with product instructions.
Description
Description
This kit comes with a separate tube of ROX reference dye. EXPRESS One-Step SYBR GreenER SuperMix kits with premixed ROX reference dye are also available.
  • Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase
  • The One-Step SuperScript™ III Reverse Transcriptase for one-step qRT-PCR
  • SYBR GreenER dye
  • UDG carryover protection - uracil-DNA-glycosylase (UDG)/dUTP to reduce carryover contamination
Order Info
Guaranteed to be stable for 6 months when properly stored.
Specifications
Specifications
EXPRESS One-Step SYBR™ GreenER™ SuperMix Kits are composed of 2 modules: EXPRESS SYBR™ GreenER™ RT module (containing SuperScript™ III Reverse Transcriptase and RNAseOUT™) and EXPRESS SYBR™ GreenER™ qPCR SuperMix with ROX in a separate tube.
Store at -20°C upon receipt. Once thawed, the EXPRESS SYBR™ GreenER™ qPCR SuperMix can be stored for up to 1 month at +4·C. Product is guaranteed to be stable for 6 months when properly stored.
Tube
Taq DNA Polymerase
7500 Fast System, 7500 System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, BioRad CFX96™, StepOnePlus™, Fast Mode, StepOnePlus™, Standard Mode, Stratagene Mx3000P, Stratagene Mx3005P, Stratagene Mx4000, BioRad MyiQ, Corbett RotorGene, Cepheid SmartCycler, MJ Opticon, MJ Chromo4, BioRad iCycler iQ
One-Step qRT-PCR Kit
500 Reactions
SYBR
1-step RT-qPCR
Fast or Standard
SYBR™ GreenER™
500 Reactions
RNA