missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EXPRESS qPCR Supermix, with premixed ROX
EXPRESS qPCR SuperMix kits are compatible with fast-cycling instruments for your probe-based qPCR assays.
£618.00 - £2565.00
Specifications
Content And Storage | EXPRESS qPCR SuperMix kits are provided as a 2X qPCR SuperMix with ROX premixed or in a separate tube. Store at -20°C upon receipt. Once thawed, the EXPRESS qPCR SuperMix can be stored for up to 1 month at +4·C. Product is guaranteedstable for 6 months when properly stored. Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). |
---|---|
Detection Method | Primer-probe |
Format | Tube |
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
10174233
|
Invitrogen™
11795200 |
200 Reactions |
£618.00
200 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10397333
|
Invitrogen™
1179501K |
1000 Reactions |
£2565.00
1000 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase
- UDG carryover protection - uracil-DNA-glycosylase (UDG)/dUTP to reduce carryover contamination
Order Info
Guaranteed minimum shelf life is 60 days.
Specifications
EXPRESS qPCR SuperMix kits are provided as a 2X qPCR SuperMix with ROX premixed or in a separate tube. Store at -20°C upon receipt. Once thawed, the EXPRESS qPCR SuperMix can be stored for up to 1 month at +4·C. Product is guaranteedstable for 6 months when properly stored. Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). |
|
Tube | |
qPCR | |
7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System, Eppendorf™ MasterCycler™ RealPlex, StepOne™, Fast Mode, StepOne™, Standard Mode | |
Gene Expression |
Primer-probe | |
High | |
Fast or Standard | |
Express qPCR Supermix | |
DNA (Genomic), cDNA |
For Research Use Only. Not for use in diagnostic procedures.