missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYBR™ GreenER™ EXPRESS SYBR™ GreenER™ qPCR Supermix, with premixed ROX
Kits are compatible with fast-cycling instruments for SYBR-based qPCR experiments.
Brand: SYBR™ GreenER™ 11794200
This item is not returnable.
View return policy
Description
This EXPRESS SYBR GreenER qPCR SuperMix comes with premixed ROX reference dye.
- Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase
- SYBR GreenER dye
- UDG carryover protection - uracil-DNA-glycosylase (UDG)/dUTP to reduce carryover contamination
Specifications
Contains 5 ml EXPRESS SYBR™ GreenER™ qPCR SuperMix with Premixed ROX. Store at −20°C for long-term storage. EXPRESS qPCR SuperMixes may be stored at 4–8°C for up to one month. | |
High | |
Taq DNA Polymerase | |
7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System, Eppendorf™ MasterCycler™ RealPlex, StepOne™, Fast Mode, StepOne™, Standard Mode | |
SYBR™ GreenER™ | |
200 reactions | |
Gene Expression | |
DNA (Genomic), cDNA |
SYBR | |
qPCR | |
Fast or Standard | |
EXPRESS SYBR GreenER qPCR SuperMix Kit | |
Real Time PCR SYBR Master Mix | |
200 Reactions | |
200 Reactions | |
5 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SYBR™ GreenER™ EXPRESS SYBR™ GreenER™ qPCR Supermix, with premixed ROX > 200 Reactions
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction