Learn More
Ecolab Extendable Handle for use with Klercide Flat Mop Frame
Extendable handle for mop frame which enables high walls or ceilings, which require mopping, to be easily reached without the health and safety issues associated with ladders or raised platforms.
Brand: Ecolab 10056567
This item is not returnable.
View return policy
Description
- Extendable handle up to 1.2 m
- Lightweight
- Autoclavable
Specifications
Autoclavable |