missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Extension Rod
PTFE construction. Extends reach by 600mm.
Brand: BRAND™ 90470
This item is not returnable.
View return policy
Description
- PTFE construction
- For use with sampling dippers
- Length: 600mm
Specifications
Extension Rod | |
PTFE | |
600 mm |