Learn More
Lenz Laborglasintrumente Extraction Thimbles Porosity 0
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05405225
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10316361
  • £47.00 / Each
Estimated Shipment: 16-08-2024
to see stock.
Add to basket
Description
Description
  • Extraction thimbles
  • Glass insert with bores for solids
  • Made of DURAN™ tubing
Specifications
Specifications
30 mL
Borosilicate Glass (3.3)
0