Learn More
Lenz Laborglasintrumente Extractor Heads Acc. To Twisselmann
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05415137
 View more versions of this product
Product Code. 10602862
  • £202.00 / Each
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Extractor heads
  • Acc. to Twisselmann
  • With vacuum jacket
  • Made of DURAN™ tubing
Specifications
Specifications
100 mL
Borosilicate Glass 3.3
1 Ea.
NS 45/40