Learn More
Lenz Laborglasintrumente Extractor Heads for Specific Heavy Solvents
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05433158
 View more versions of this product
Product Code. 10661702
  • £373.00 / Each
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For the extraction of liquids with specific heavy solvents
  • With insert filter (porosity: 0)
  • Made of DURAN™ tubing
Specifications
Specifications
500 mL
Fritte P0
NS 60/46